Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011530.3 | NP_035660.3 | 7, 8 | 1433 | ||
GenBank mRNA | AK020022.1 | - | 6, 7 | 1509 | ||
AK079667.1 | - | 7, 8 | 1347 | |||
AK145234.1 | - | 7, 8 | 1393 | |||
AK151376.1 | - | 7, 8 | 1343 | |||
AK151483.1 | - | 7, 8 | 1343 | |||
AK151657.1 | - | 7, 8 | 1366 | |||
AK153287.1 | - | 7, 8 | 1343 | |||
AK159200.1 | - | 7, 8 | 1399 | |||
AK161091.1 | - | 7, 8 | 1349 | |||
BC005578.1 | - | 7, 8 | 1333 | 3968225 | ||
BC051257.1 | - | 7, 8 | 1346 | 6530488 | ||
BC057668.1 | - | 7, 8 | 1720 | |||
CT010387.1 | - | 6, 7 | 1280 | |||
M90459.1 | - | 6, 7 | 1280 | |||
U60087.1 | - | 6, 7 | 1283 | |||
U60089.1 | - | 6, 7 | 1283 | |||
U60090.1 | - | 6, 7 | 1283 | |||
U60091.1 | - | 6, 7 | 1283 | |||
U60092.1 | - | 6, 7 | 1280 |