Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159750.1 | NP_001153222.1 | 10 | 1627 | ||
NM_001159751.1 | NP_001153223.1 | 10 | 1523 | |||
NM_011541.4 | NP_035671.1 | 10 | 1630 | |||
GenBank mRNA | AK136035.1 | - | 10 | 1523 | ||
AK143593.1 | - | 10 | 1488 | |||
BC006022.1 | - | 10 | 1504 | 3590733 | ||
BC061490.1 | - | 10 | 1522 | 2582185 | ||
BC083127.1 | - | 10 | 1493 | 6390364 | ||
M18209.1 | - | 9 | 1428 |