Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011543.4 | NP_035673.3 | 4 | 372 | ||
GenBank mRNA | AF083214.1 | - | 4 | 296 | ||
AK002983.1 | - | 4 | 343 | |||
AK010628.1 | - | 4 | 358 | |||
AK012498.1 | - | 4 | 331 | |||
AK014245.1 | - | 4 | 331 | |||
AK027909.1 | - | 4 | 330 | |||
AK078330.1 | - | 4 | 358 | |||
AK088339.1 | - | 4 | 331 | |||
AK152609.1 | - | 4 | 330 | |||
AK160198.1 | - | 4 | 331 | |||
AK166673.1 | - | 4 | 331 | |||
BC002115.1 | - | 4 | 315 | 3490159 | ||
BU841654.1 | - | 5 | 464 | |||
Z47088.1 | - | 4 | 321 |