Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011549.3 | NP_035679.3 | 1 | 153 | ||
GenBank mRNA | AF079095.1 | - | 1 | 134 | ||
AK146016.1 | - | 1 | 178 | |||
BC064789.1 | - | 1 | 153 | 30429545 |