Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098226.3 | NP_001091696.2 | 13 | 2228 | ||
NM_001204156.1 | NP_001191085.1 | 13 | 2551 | |||
NM_011566.4 | NP_035696.3 | 13 | 2392 | |||
GenBank mRNA | AF002670.1 | - | 13 | 2347 | ||
AK087649.1 | - | 13 | 2232 | |||
AK143551.1 | - | 12 | 3262 | |||
BC021364.1 | - | 13 | 2272 | |||
BC027383.1 | - | 4 | 1303 | |||
BC039984.1 | - | 4 | 1338 | |||
BC047929.1 | - | 13 | 2551 | |||
D87963.1 | - | 12 | 2062 | |||
Y10027.1 | - | 12 | 2160 |