Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011574.2 | NP_035704.2 | 6 | 701 | ||
GenBank mRNA | AK144455.1 | - | 6 | 686 | ||
AK146263.1 | - | 6 | 677 | |||
AK150862.1 | - | 6 | 676 | |||
AK168470.1 | - | 6 | 651 | |||
AK173318.1 | - | 5 | 642 | |||
AK178383.1 | - | 2 | 150 | |||
AK178714.1 | - | 2 | 150 | |||
AK187526.1 | - | 2 | 150 | |||
AK195627.1 | - | 2 | 155 | |||
AK208986.1 | - | 2 | 150 | |||
BC027399.1 | - | 2 | 200 | 4948207 | ||
BC068113.1 | - | 6 | 690 | 5707865 |