Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011574.2 | NP_035704.2 | 13 | 1610 | ||
GenBank mRNA | AK144455.1 | - | 13 | 1595 | ||
AK146263.1 | - | 13 | 1586 | |||
AK150862.1 | - | 13 | 1585 | |||
AK168470.1 | - | 13 | 1560 | |||
AK173318.1 | - | 12 | 1551 | |||
BC018262.1 | - | 1 | 63 | |||
BC027399.1 | - | 9 | 1109 | 4948207 | ||
BC068113.1 | - | 13 | 1599 | 5707865 |