Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011578.3 | NP_035708.2 | 5 | 897 | ||
GenBank mRNA | AF039601.1 | - | 5 | 697 | ||
AK133234.1 | - | 3 | 405 | |||
AK165763.1 | - | 5 | 813 | |||
AK166082.1 | - | 5 | 898 | |||
BC070428.1 | - | 5 | 822 | 6410554 |