Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011580.3 | NP_035710.2 | 5 | 1078 | ||
GenBank mRNA | AK080686.1 | - | 5 | 1113 | ||
AK145202.1 | - | 5 | 1108 | |||
AK163092.1 | - | 5 | 1105 | |||
BC020435.1 | - | 5 | 1086 | |||
BC042422.1 | - | 5 | 1080 | 4018176 | ||
BC050917.1 | - | 5 | 1078 | 6405710 | ||
M87276.1 | - | 5 | 1104 |