Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tie1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011587.2 | NP_035717.2 | 5 | 849 | ||
GenBank mRNA | AK051512.1 | - | 5 | 795 | ||
AK052192.1 | - | 5 | 769 | |||
AK052413.1 | - | 5 | 782 | |||
AK161898.1 | - | 5 | 768 | |||
BC046452.1 | - | 5 | 771 | 4195879 | ||
BC057004.1 | - | 5 | 755 | 6831929 | ||
BC060182.1 | - | 5 | 755 | 6831929 | ||
X71425.1 | - | 5 | 750 | |||
X73960.1 | - | 5 | 749 | |||
X80764.1 | - | 5 | 1063 |