Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011598.3 | NP_035728.2 | 4 | 620 | ||
GenBank mRNA | AK005745.1 | - | 4 | 564 | ||
AK161314.1 | - | 4 | 525 | |||
BC048437.1 | - | 4 | 559 | 6773242 | ||
CN837748.1 | - | 331 |