Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF229633.1 | - | 13 | 1257 | ||
AK122481.2 | - | 11 | 1116 | |||
AK134989.1 | - | 13 | 1891 | |||
BC086631.1 | - | 14 | 1801 | |||
BU149633.1 | - | 295 | ||||
U61363.1 | - | 5 | 660 |