Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF229633.1 | - | 20 | 2340 | ||
AK122481.2 | - | 18 | 2199 | |||
BC019750.1 | - | 4 | 405 | |||
BC043477.1 | - | 5 | 554 | |||
BC058525.1 | - | 3 | 293 | |||
BC086631.1 | - | 21 | 2883 | |||
BC086780.1 | - | 3 | 233 | |||
CB058377.1 | - | 3 | 247 | |||
U61363.1 | - | 12 | 1743 |