Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011605.3 | NP_035735.2 | 4 | 1258 | ||
GenBank mRNA | AK144642.1 | - | 4 | 1218 | ||
AK156206.1 | - | 3 | 1889 | |||
BC141062.1 | - | 4 | 976 | 9055674 | ||
U39078.1 | - | 4 | 1172 |