Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130174.2 | NP_001123646.1 | 6, 7 | 309 | ||
NM_001130175.2 | NP_001123647.4 | 6, 7 | 305 | |||
NM_001130176.1 | NP_001123648.1 | 4, 5 | 196 | |||
NM_001130177.2 | NP_001123649.1 | 5, 6 | 297 | |||
NM_001130178.2 | NP_001123650.1 | 5, 6 | 279 | |||
NM_001130179.2 | NP_001123651.4 | 5, 6 | 275 | |||
NM_001130180.2 | NP_001123652.1 | 4, 5 | 267 | |||
NM_001130181.2 | NP_001123653.4 | 4, 5 | 263 | |||
NM_011619.3 | NP_035749.1 | 4, 5 | 267 | |||
GenBank mRNA | AK146945.1 | - | 6, 7 | 254 | ||
AK163786.1 | - | 5, 6 | 242 | |||
AK168393.1 | - | 5, 6 | 242 | |||
AK169061.1 | - | 5, 6 | 224 | |||
BC063753.1 | - | 6, 7 | 285 | 30293959 | ||
BQ554639.1 | - | 5, 6 | 215 | |||
L47549.1 | - | 5, 6 | 193 | |||
L47550.1 | - | 5, 6 | 178 | |||
L47551.1 | - | 4, 5 | 179 | |||
L47552.1 | - | 4, 5 | 188 | |||
L47553.1 | - | 4, 5 | 195 | |||
L47599.1 | - | 5, 6 | 199 | |||
L47600.1 | - | 4, 5 | 196 |