Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011624.2 | NP_035754.1 | 8 | 1068 | ||
GenBank mRNA | AB013603.1 | - | 7 | 881 | ||
AK041159.1 | - | 8 | 978 | |||
AK144709.1 | - | 9 | 1054 | |||
AK146475.1 | - | 8 | 1090 | |||
AK163796.1 | - | 1 | 77 | |||
AK205914.1 | - | 1 | 61 | |||
BC006614.1 | - | 6 | 2355 | |||
BC031723.1 | - | 8 | 1068 | 5003002 |