Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011629.3 | NP_035759.3 | 11 | 1442 | ||
GenBank mRNA | AK146891.1 | - | 11 | 1455 | ||
AK149374.1 | - | 11 | 1357 | |||
AK212545.1 | - | 2 | 66 | |||
BC090662.1 | - | 11 | 1418 | 30682075 | ||
BC094580.1 | - | 11 | 1360 | 30602939 | ||
L26957.1 | - | 10 | 1251 | |||
U28265.1 | - | 10 | 1251 | |||
U30482.1 | - | 10 | 1251 | |||
Y11436.1 | - | 10 | 1890 |