Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011632.3 | NP_035762.2 | 8 | 1055 | ||
GenBank mRNA | AK148231.1 | - | 2 | 211 | ||
BC137634.1 | - | 7 | 770 | 8860653 | ||
BC137635.1 | - | 7 | 770 | 8860654 | ||
U21050.1 | - | 8 | 874 |