Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286122.1 | NP_001273051.1 | 9 | 1269 | ||
NM_011632.3 | NP_035762.2 | 11 | 1455 | |||
GenBank mRNA | AK147304.1 | - | 9 | 1197 | ||
AK147362.1 | - | 9 | 1270 | |||
AK148231.1 | - | 5 | 610 | |||
BC137634.1 | - | 10 | 1170 | 8860653 | ||
BC137635.1 | - | 10 | 1170 | 8860654 | ||
U21050.1 | - | 11 | 1274 |