Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011634.3 | NP_035764.2 | 15 | 1997 | ||
GenBank mRNA | AK012948.1 | - | 15 | 1996 |