Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011639.3 | NP_035769.1 | 6 | 1084 | ||
GenBank mRNA | AF097511.1 | - | 6 | 977 | ||
AF116823.1 | - | 6 | 1079 | |||
AK010435.1 | - | 6 | 1085 | |||
AK146823.1 | - | 6 | 1034 | |||
AK159387.1 | - | 6 | 1053 | |||
AK166024.1 | - | 6 | 1248 | |||
BC138685.1 | - | 6 | 1016 | 8861706 | ||
BC138686.1 | - | 6 | 1016 | 8861707 |