Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127259.1 | NP_001120731.1 | 14 | 2887 | ||
NM_001127260.1 | NP_001120732.1 | 13 | 2793 | |||
NM_001127262.1 | NP_001120734.1 | 11 | 2130 | |||
NM_001127264.1 | NP_001120736.1 | 12 | 2212 | |||
NM_011641.2 | NP_035771.1 | 12 | 2224 | |||
GenBank mRNA | AB010152.1 | - | 12 | 2199 | ||
AK137253.1 | - | 12 | 2213 | |||
BC090649.1 | - | 12 | 2194 | 30645427 |