Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126330.1 | NP_001119802.1 | 3 | 696 | ||
NM_001126331.1 | NP_001119803.1 | 3 | 696 | |||
NM_011642.3 | NP_035772.2 | 5 | 986 | |||
GenBank mRNA | AB221594.1 | - | 4 | 578 | ||
AK017412.1 | - | 2 | 227 | |||
AK139633.1 | - | 3 | 697 | |||
BC066045.1 | - | 3 | 583 | 6812399 | ||
Y19234.1 | - | 5 | 987 | |||
Y19235.1 | - | 3 | 690 |