Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011653.2 | NP_035783.1 | 4 | 1560 | ||
GenBank mRNA | AK077529.1 | - | 4 | 1571 | ||
AK145404.1 | - | 4 | 1609 | |||
AK146983.1 | - | 4 | 1609 | |||
AK161783.1 | - | 2 | 1156 | |||
AK164335.1 | - | 4 | 1611 | |||
AK167687.1 | - | 4 | 1609 | |||
AK168762.1 | - | 4 | 1609 | |||
AK169610.1 | - | 4 | 1610 | |||
AK199768.1 | - | 1 | 325 | |||
AK216974.1 | - | 1 | 357 | |||
BC056169.1 | - | 4 | 1571 | 5715646 | ||
BC083343.1 | - | 4 | 1584 | 5687414 | ||
BC083344.1 | - | 4 | 1577 | 5693701 | ||
BC083345.1 | - | 4 | 1594 | 6415067 | ||
BC085256.1 | - | 4 | 1593 | 6306481 | ||
M13445.1 | - | 4 | 1562 | |||
M28729.1 | - | 1 | 755 |