Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011659.2 | NP_035789.1 | 4, 5 | 615 | ||
GenBank mRNA | BC065782.1 | - | 4, 5 | 540 | ||
BC139239.1 | - | 4, 5 | 485 | 8862261 | ||
BC139266.1 | - | 4, 5 | 485 | 8862288 | ||
Z21674.1 | - | 4, 5 | 436 |