Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011662.2 | NP_035792.1 | 5 | 513 | ||
GenBank mRNA | AK154480.1 | - | 5 | 512 | ||
BC056450.1 | - | 5 | 513 | 6814478 |