Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011669.3 | NP_035799.1 | 9 | 2200 | ||
GenBank mRNA | AK035629.1 | - | 9 | 2041 | ||
AK167153.1 | - | 9 | 2040 | |||
BC023858.1 | - | 3 | 1033 | |||
BC049274.1 | - | 7 | 1867 | 4500832 | ||
BC055398.1 | - | 5 | 1292 | |||
BC068136.1 | - | 9 | 2200 | 30363916 |