Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 3 | 387 | ||
GenBank mRNA | AK138838.1 | - | 3 | 322 | ||
AK143582.1 | - | 3 | 322 | |||
AK149964.1 | - | 3 | 345 | |||
AK169372.1 | - | 3 | 366 | |||
AK169933.1 | - | 3 | 357 | |||
AK171271.1 | - | 3 | 357 | |||
AK214431.1 | - | 2 | 183 | |||
AK218565.1 | - | 2 | 183 | |||
AK220512.1 | - | 3 | 344 | |||
BC011341.1 | - | 3 | 305 | 3661165 | ||
BC066180.1 | - | 3 | 297 | |||
BC066865.1 | - | 3 | 297 | |||
L00681.1 | - | 3 | 350 |