Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011682.4 | NP_035812.3 | 4 | 354 | ||
GenBank mRNA | AK049602.1 | - | 5 | 711 | ||
AK134010.1 | - | 4 | 348 | |||
AK134424.1 | - | 5 | 966 | |||
AK163051.1 | - | 4 | 357 | |||
BC011062.1 | - | 5 | 403 | |||
BC050183.1 | - | 5 | 463 | |||
BC062163.1 | - | 5 | 410 | |||
Y12229.1 | - | 4 | 265 |