Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 7 | 535 | ||
GenBank mRNA | AK012359.1 | - | 7 | 533 | ||
AK078555.1 | - | 7 | 539 | |||
AK150504.1 | - | 7 | 561 | |||
AK150627.1 | - | 7 | 561 | |||
AK150940.1 | - | 7 | 553 | |||
AK152274.1 | - | 7 | 533 | |||
AK159561.1 | - | 7 | 770 | |||
AK168199.1 | - | 7 | 506 | |||
AK168328.1 | - | 7 | 532 | |||
BC003731.1 | - | 7 | 535 | 3601668 | ||
U30838.1 | - | 7 | 535 |