Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011695.2 | NP_035825.1 | 10 | 1423 | ||
GenBank mRNA | AK078555.1 | - | 10 | 1429 | ||
AK150940.1 | - | 10 | 1441 | |||
AK168328.1 | - | 10 | 1420 | |||
U30838.1 | - | 10 | 1423 |