Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011699.3 | NP_035829.1 | 5 | 468 | ||
GenBank mRNA | AF087695.1 | - | 5 | 468 | ||
AF173083.1 | - | 5 | 475 | |||
AK015399.1 | - | 5 | 487 | |||
AK078923.1 | - | 5 | 488 | |||
AK083299.1 | - | 5 | 492 | |||
AK128989.1 | - | 5 | 488 | |||
AK135810.1 | - | 6 | 494 | |||
AK161076.1 | - | 5 | 449 | |||
AK162185.1 | - | 3 | 313 | |||
BC046966.1 | - | 5 | 468 | 6493071 | ||
BC052471.1 | - | 5 | 478 |