Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011699.3 | NP_035829.1 | 5 | 1739 | ||
GenBank mRNA | AK078213.1 | - | 1 | 467 | ||
AK088361.1 | - | 1 | 536 | |||
AK128989.1 | - | 5 | 1759 | |||
AK161076.1 | - | 5 | 1720 | |||
BC046966.1 | - | 5 | 1739 | 6493071 | ||
BC052471.1 | - | 5 | 1749 |