Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164567.1 | NP_001158039.1 | 10 | 1028 | ||
NM_011700.2 | NP_035830.2 | 10 | 1028 | |||
GenBank mRNA | AJ344341.1 | - | 10 | 1016 | ||
AK028229.1 | - | 10 | 1029 | |||
AK166823.1 | - | 9 | 1260 | |||
BC021808.1 | - | 10 | 1004 | 3991439 | ||
BQ833192.1 | - | 2 | 99 | |||
CF171710.1 | - | 6 | 585 |