Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164567.1 | NP_001158039.1 | 12 | 1262 | ||
NM_011700.2 | NP_035830.2 | 12 | 1262 | |||
GenBank mRNA | AJ344341.1 | - | 12 | 1250 | ||
AK028229.1 | - | 12 | 1263 | |||
AK166652.1 | - | 2 | 176 | |||
AK166823.1 | - | 11 | 1494 | |||
BC021808.1 | - | 12 | 1238 | 3991439 | ||
BC022664.1 | - | 3 | 191 | 4223317 |