Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011701.4 | NP_035831.2 | 3 | 793 | ||
GenBank mRNA | AK012469.1 | - | 3 | 296 | ||
AK131618.1 | - | 3 | 795 | |||
AK133726.1 | - | 3 | 794 | |||
AK153019.1 | - | 3 | 796 | |||
AK159539.1 | - | 3 | 794 | |||
AK169181.1 | - | 3 | 795 | |||
AK170429.1 | - | 3 | 794 | |||
AK198215.1 | - | 1 | 24 | |||
AK208074.1 | - | 1 | 24 | |||
AK209588.1 | - | 1 | 24 | |||
AK209779.1 | - | 1 | 24 | |||
AK210390.1 | - | 1 | 24 | |||
AK210884.1 | - | 1 | 24 | |||
AK212337.1 | - | 1 | 24 | |||
AK212338.1 | - | 1 | 24 | |||
AK212907.1 | - | 1 | 24 | |||
AK213561.1 | - | 1 | 24 | |||
AK216501.1 | - | 1 | 24 | |||
AK217097.1 | - | 1 | 24 | |||
AK218318.1 | - | 1 | 24 | |||
BC089335.1 | - | 3 | 789 | 6415629 | ||
M24849.1 | - | 3 | 710 | |||
M26251.1 | - | 3 | 1152 | |||
X51438.1 | - | 3 | 762 | |||
X56397.1 | - | 3 | 756 |