Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011702.2 | NP_035832.1 | 7 | 1109 | ||
GenBank mRNA | AK018599.1 | - | 7 | 1110 | ||
BC089511.1 | - | 7 | 1016 | 30249277 | ||
Z31361.1 | - | 1 | 104 |