Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029843.1 | NP_001025014.1 | 7, 8 | 685 | ||
NM_001029844.1 | NP_001025015.1 | 7, 8 | 685 | |||
NM_011705.3 | NP_035835.1 | 7, 8 | 685 | |||
GenBank mRNA | AF080252.1 | - | 6, 7 | 598 | ||
AF080253.1 | - | 7, 8 | 639 | |||
AK032344.1 | - | 7, 8 | 656 | |||
AK039554.1 | - | 1, 2 | 57 | |||
AK088230.1 | - | 7, 8 | 609 | |||
AK088674.1 | - | 7, 8 | 668 | |||
AK136352.1 | - | 7, 8 | 659 | |||
AK166676.1 | - | 7, 8 | 653 | |||
BC016676.1 | - | 7, 8 | 720 | 3602292 | ||
BC051072.1 | - | 7, 8 | 665 |