Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011706.2 | NP_035836.2 | 5 | 927 | ||
GenBank mRNA | AB021665.1 | - | 5 | 1009 | ||
AK008689.1 | - | 3 | 452 | |||
AK089004.1 | - | 6 | 942 | |||
AK134052.1 | - | 4 | 1240 | |||
AK162841.1 | - | 4 | 801 | |||
AK171222.1 | - | 4 | 850 | |||
BC005415.1 | - | 4 | 874 | 3487527 |