Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164314.1 | NP_001157786.1 | 3 | 430 | ||
NM_001164488.1 | NP_001157960.1 | 3 | 363 | |||
NM_011710.3 | NP_035840.3 | 3 | 376 | |||
GenBank mRNA | AK004541.1 | - | 3 | 364 | ||
AK150090.1 | - | 3 | 394 | |||
AK150267.1 | - | 4 | 478 | |||
AK152966.1 | - | 3 | 394 | |||
AK153337.1 | - | 3 | 402 | |||
BC003450.1 | - | 3 | 362 | 2654231 | ||
BC046232.1 | - | 3 | 369 | 6813406 | ||
CD563585.1 | - | 3 | 350 | |||
CT010258.1 | - | 2 | 262 |