Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011711.1 | NP_035841.1 | 4 | 484 | ||
GenBank mRNA | AK129473.1 | - | 4 | 403 | ||
AK139711.1 | - | 4 | 467 | |||
AK143308.1 | - | 4 | 383 | |||
AK155331.1 | - | 4 | 501 | |||
AK163361.1 | - | 4 | 468 | |||
AK170744.1 | - | 4 | 467 | |||
BC023797.1 | - | 3 | 204 | |||
BC085094.1 | - | 4 | 381 | |||
BC131961.1 | - | 4 | 360 | 40130238 |