Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011729.2 | NP_035859.2 | 6 | 971 | ||
GenBank mRNA | AK038822.1 | - | 6 | 800 | ||
AK137563.1 | - | 6 | 803 | |||
AK140825.1 | - | 6 | 800 | |||
AK160645.1 | - | 4 | 343 | |||
AK164205.1 | - | 6 | 789 | |||
BC033413.1 | - | 6 | 691 | |||
BC053034.1 | - | 6 | 780 | |||
BC060659.1 | - | 6 | 770 | |||
BC092235.1 | - | 6 | 762 | |||
BC141070.1 | - | 6 | 801 | 9055682 | ||
D16306.1 | - | 6 | 666 | |||
U40796.1 | - | 6 | 867 |