Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011738.2 | NP_035868.1 | 2 | 366 | ||
GenBank mRNA | AF077002.1 | - | 2 | 291 | ||
AK077596.1 | - | 2 | 229 | |||
AK149224.1 | - | 2 | 336 | |||
AK166922.1 | - | 2 | 165 | |||
AK168520.1 | - | 2 | 228 | |||
AK169035.1 | - | 2 | 217 | |||
AK169189.1 | - | 2 | 165 | |||
AK187410.1 | - | 2 | 65 | |||
BC008187.1 | - | 2 | 155 | 3589806 | ||
BC061497.1 | - | 2 | 310 | 4949445 | ||
CB183421.1 | - | 2 | 366 | |||
D87661.1 | - | 2 | 298 | |||
U57311.1 | - | 2 | 114 |