Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291485.1 | NP_001278414.1 | 2 | 262 | ||
NM_006890.4 | NP_008821.2 | 2 | 262 | |||
GenBank mRNA | AF006622.1 | - | 2 | 153 | ||
AF006623.1 | - | 2 | 153 | |||
AK292213.1 | - | 2 | 255 | |||
AK296634.1 | - | 2 | 255 | |||
AK311285.1 | - | 2 | 90 | |||
AK313108.1 | - | 2 | 255 | |||
BC121132.2 | - | 2 | 156 | 40120888 | ||
DC345133.1 | - | 2 | 262 | |||
L31792.1 | - | 2 | 152 | |||
X98311.1 | - | 2 | 152 |