Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110791.1 | NP_001104261.1 | 4 | 659 | ||
NM_011750.2 | NP_035880.2 | 4 | 659 | |||
GenBank mRNA | AK133895.1 | - | 4 | 448 | ||
AK146151.1 | - | 4 | 420 | |||
AK147081.1 | - | 4 | 428 | |||
AK152914.1 | - | 3 | 361 | |||
AK154705.1 | - | 4 | 428 | |||
AK157845.1 | - | 4 | 428 | |||
AK214037.1 | - | 3 | 236 | |||
BC009091.1 | - | 4 | 404 | 3157495 | ||
BC055370.1 | - | 4 | 418 | 30055109 | ||
X85802.1 | - | 4 | 416 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U50960.1 | - | 19 | Chr. - - [-] on Build |