Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011752.2 | NP_035882.1 | 14 | 1722 | ||
GenBank mRNA | AB041612.1 | - | 14 | 1605 | ||
AK016551.1 | - | 8 | 889 | |||
AK158995.1 | - | 14 | 1683 | |||
AK167849.1 | - | 14 | 1593 | |||
AK168061.1 | - | 14 | 1564 | |||
AK170372.1 | - | 5 | 2385 | |||
BC021397.1 | - | 14 | 1605 | 5067274 |