Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011755.2 | NP_035885.2 | 3 | 660 | ||
GenBank mRNA | AK132703.1 | - | 3 | 663 | ||
BC138830.1 | - | 3 | 579 | 8861852 | ||
BC138832.1 | - | 2 | 577 | 8861854 | ||
X17617.1 | - | 3 | 611 |