Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044703.2 | NP_001038168.1 | 2 | 258 | ||
NM_001044704.2 | NP_001038169.1 | 3 | 324 | |||
NM_001044705.2 | NP_001038170.1 | 2 | 239 | |||
NM_011757.3 | NP_035887.2 | 3 | 343 | |||
GenBank mRNA | AK146776.1 | - | 3 | 317 | ||
AK146875.1 | - | 3 | 304 | |||
BC010591.1 | - | 2 | 186 | 3981599 | ||
BU513148.1 | - | 2 | 200 | |||
BY034027.1 | - | 3 | 327 | |||
BY293243.1 | - | 3 | 325 | |||
BY309906.1 | - | 2 | 226 | |||
CF915830.1 | - | 3 | 268 | |||
D10630.1 | - | 2 | 177 | |||
DV651309.1 | - | 3 | 356 | |||
U41671.1 | - | 3 | 274 | |||
X63747.1 | - | 2 | 1034 |