Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011775.6 | NP_035905.1 | 5 | 397 | ||
GenBank mRNA | AK135903.1 | - | 5 | 397 | ||
AK138699.1 | - | 5 | 398 | |||
BC071183.1 | - | 5 | 379 | 30473497 | ||
M34148.1 | - | 5 | 397 |