Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289617.1 | NP_001276546.1 | 5 | 1158 | ||
NM_001289618.1 | NP_001276547.1 | 4 | 1065 | |||
NM_001289619.1 | NP_001276548.1 | 4 | 1065 | |||
NM_011777.3 | NP_035907.1 | 5 | 1158 | |||
GenBank mRNA | AK031415.1 | - | 5 | 956 | ||
AK035743.1 | - | 5 | 929 | |||
AK076491.1 | - | 5 | 928 | |||
AK090110.1 | - | 5 | 925 | |||
AK147812.1 | - | 5 | 1159 | |||
AK161709.1 | - | 4 | 860 | |||
AK165290.1 | - | 3 | 999 | |||
AK170571.1 | - | 5 | 932 | |||
AK176971.1 | - | 1 | 218 | |||
BC054775.1 | - | 4 | 1024 | 6808149 | ||
Y07711.1 | - | 4 | 866 |